Få mere at vide
Tegaserod Maleate, TRC
Artikelnummer. 30423698
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423698 100 mg
30425832 25 mg
30423241 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423698 Leverandør Toronto Research Chemicals Leverandørnr. TRCT014500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 189188-57-6
Molekylær formel C16 H23 N5 O . C4 H4 O4
Molekylvægt (g/mol) 417.46
Synonym Hydrazinecarboximidamide, 2-[(5-methoxy-1H-indol-3-yl)methylene]-N-pentyl-, (2Z)-2-butenedioate (1:1), Hydrazinecarboximidamide, 2-[(5-methoxy-1H-indol-3-yl)methylene]-N-pentyl-, (Z)-2-butenedioate (1:1), HTF 919, SDZ-HTF 919, Tegaserod hydrogen maleate, Tegaserod maleate, Tegibs 6, Zelmac, Zelnorm
IUPAC navn (Z)-but-2-enedioic acid;1-[(5-methoxy-1H-indol-3-yl)methylideneamino]-3-pentylguanidine
SMIL CCCCCNC(=N)NN=Cc1c[nH]c2ccc(OC)cc12.OC(=O)\C=C/C(=O)O
Tekniske data
Kemisk navn eller materiale Tegaserod, Maleate
Mængde 100 mg
InChI formel InChI=1S/C16H23N5O.C4H4O4/c1-3-4-5-8-18-16(17)21-20-11-12-10-19-15-7-6-13(22-2)9-14(12)15;5-3(6)1-2-4(7)8/h6-7,9-11,19H,3-5,8H2,1-2H3,(H3,17,18,21);1-2H,(H,5,6)(H,7,8)/b;2-1-
Formel vægt 417.2012