Learn More
Macherey-Nagel™ Tools for Valco Fused Silica Adapters
Work easily with one-piece and removable FS adapters. Macherey-Nagel™ Tools for Valco Fused Silica Adapters include an open end wrench (3/16 and 1/4 in.) for 1/32 in. fittings and a pin vise/drill index set (0.34 - 1.0mm).
Brand: Macherey-Nagel™ 724423
This item is not returnable.
View return policy
Description
- Open end wrench: 3/16 x 1/4 in. size, for use with 1/32 in. fittings
- Pin vise and drill index: 0.34 - 1.0mm
Specifications
Open End Wrench | |
0.34 to 1.0 mm | |
Capillary Gas Chromatography | |
1 Each |
Product Content Correction
Product Title
Macherey-Nagel™ Tools for Valco Fused Silica Adapters > Type: Open End Wrench
Spot an opportunity for improvement?Share a Content Correction