Få mere at vide
Tedizolid Phosphate, TRC
Artikelnummer. 30426289
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30426289 1 mg
30423501 5 mg
30422006 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30426289 Leverandør Toronto Research Chemicals Leverandørnr. TRCT0137551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 856867-55-5
Molekylær formel C17 H16 F N6 O6 P
Molekylvægt (g/mol) 450.32
Synonym (5R)-3-[3-Fluoro-4-[6-(2-methyl-2H-tetrazol-5-yl)-3-pyridinyl]phenyl]-5-[(phosphonooxy)methyl]-2-oxazolidinone, TR 701FA, Tedizolid Hydrogen Phosphate, Tedizolid Phosphate
IUPAC navn [(5R)-3-[3-fluoro-4-[6-(2-methyltetrazol-5-yl)pyridin-3-yl]phenyl]-2-oxo-1,3-oxazolidin-5-yl]methyl dihydrogen phosphate
SMIL Cn1nnc(n1)c2ccc(cn2)c3ccc(cc3F)N4C[C@H](COP(=O)(O)O)OC4=O
Tekniske data
Kemisk navn eller materiale Tedizolid Phosphate
Mængde 1 mg
InChI formel InChI=1S/C17H16FN6O6P/c1-23-21-16(20-22-23)15-5-2-10(7-19-15)13-4-3-11(6-14(13)18)24-8-12(30-17(24)25)9-29-31(26,27)28/h2-7,12H,8-9H2,1H3,(H2,26,27,28)/t12-/m1/s1
Formel vægt 450.0853