Få mere at vide
BRAND™ Technical Quality PFA Bottle
Shop alle Brand produkter
Kapacitet (metrisk):
100 mL
500 mL
Pakningsstørrelse:
Hver
Beskrivelse
- With screw caps made of ETFE
- These bottles contain a small fraction of reprocessed PFA from our production process
- The physical properties and chemical resistance compare with high purity PFA
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 500 mL |
| Materiale | PFA |
| Mund | Narrow |
| Lukningsmateriale | ETFE |
| Lukketype | Screw Cap |