missing translation for 'onlineSavingsMsg'
Learn More

Techne™ Filter Cartridges for PrimeQ Real-time PCR System

Incorporates both excitation and emission filters together with dichroic mirror for fluorophore excitation and emission detection

Brand:  Techne™ FC01

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15490340

  • 7454.97 DKK / Each


Estimated Shipment
11-11-2020
Add to basket

Description

Description

  • Optimized for use with FAM™, HEX™, ROX™ and Cy™5 dyes
  • Fluorescence excitation range : 470-650nm (standard filters are centered at 485, 530, 580 and 640nm)
  • Fluorescence detection range : 500-710nm (standard filters are centered at 520, 560, 615 and 685nm)
  • FC02, FC03, FC04 and FC05 filter cartridges are supplied as standard with PrimeQ system
  • FC01 is not included with unit

Specifications

Specifications

FAM multiplex, green intercalating dyes, Fluorescein, EvaGreen
460λ
Filter Cartridge
500λ
Videos
Documents