missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tebufenozide, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Tebufenozide |
| CAS | 112410-23-8 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C22 H28 N2 O2 |
| Mængde | 100 mg |
| Synonym | Benzoic acid','3,5-dimethyl-','1-(1,1-dimethylethyl)-2-(4-ethylbenzoyl)hydrazide (9CI','ACI), 1-tert-Butyl-1-(3,5-dimethylbenzoyl)-2-(4-ethylbenzoyl)hydrazine, 3,5-Dimethylbenzoic acid N-tert-butyl-N-(4-ethylbenzoyl)hydrazide, Confirm, Mimic, Mimic 240LV, Mimic 700WP, RH 5992, Romdan, Tebufenozide |
| InChI formel | InChI=1S/C22H28N2O2/c1-7-17-8-10-18(11-9-17)20(25)23-24(22(4,5)6)21(26)19-13-15(2)12-16(3)14-19/h8-14H,7H2,1-6H3,(H,23,25) |
| SMIL | CCc1ccc(cc1)C(=O)NN(C(=O)c2cc(C)cc(C)c2)C(C)(C)C |
| IUPAC navn | N-tert-butyl-N'-(4-ethylbenzoyl)-3,5-dimethylbenzohydrazide |
| Molekylvægt (g/mol) | 352.47 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?