Få mere at vide
Tebuconazole-d9, TRC
Artikelnummer. 30419796
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30419796 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30419796 Leverandør Toronto Research Chemicals Leverandørnr. TRCT01300210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tebuconazole-d9
CAS 1246818-83-6
Renhedsgrad noter HPLC
Molekylær formel C16H13D9ClN3O
Mængde 10 mg
Synonym Tebuconazole-d9, Tebuconazole-D9 (tert-butyl-D9), 5-(4-Chlorophenyl)-1,1,1-trideuterio-3-(1,2,4-triazol-1-ylmethyl)-2,2-bis(trideuteriomethyl)pentan-3-ol, α-[2-(4-Chlorophenyl)ethyl]-α-[1,1-di(methyl-d3)ethyl-2,2,2-d3]-1H-1,2,4-triazole-1-ethanol, 1-(4-Chlorophenyl)-4,4-di(methyl-d3)-3-(1,2,4-triazol-1-yl-methyl)pentan-3-ol-5,5,5-d3, Tebuconazole D9 (trimethyl D9)
InChI formel InChI=1S/C16H22ClN3O/c1-15(2,3)16(21,10-20-12-18-11-19-20)9-8-13-4-6-14(17)7-5-13/h4-7,11-12,21H,8-10H2,1-3H3/i1D3,2D3,3D3
SMIL OC(CN1C=NC=N1)(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])CCC2=CC=C(Cl)C=C2
Molekylvægt (g/mol) 316.87
Formel vægt 316.2016
Procent renhed >95