missing translation for 'onlineSavingsMsg'
Learn More
Tebipenem, MedChemExpress Product Code.: 16488288

Tebipenem, MedChemExpress

Product Code. p-200098422
10 mg, 10mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488288

Brand: Medchem Express HYA0076

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tebipenem is an orally available carbapenem antibiotic, shows broad-spectrum activity against Gram-positive and -negative bacteria, except for Pseudomonas aeruginosa.

Tebipenem is an orally available carbapenem antibiotic, shows broad-spectrum activity against Gram-positive and -negative bacteria, except for Pseudomonas aeruginosa.

Specifications

Chemical Name or Material Tebipenem
Physical Form Solid
CAS 161715-21-5
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H21N3O4S2
For Use With (Application) COVID-19-immunoregulation
Synonym LJC 11036
Solubility Information DMSO : 33.33 mg/mL (86.91 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (18.62 mM; Need ultrasonic)
SMILES O=C(C(N1C2=O)=C(SC3CN(C4=NCCS4)C3)[C@H](C)[C@]1([H])[C@@]2([H])[C@H](O)C)O
Molecular Weight (g/mol) 383.49
Formula Weight 383.49
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Tebipenem, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.