missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ TCH-165
GREENER_CHOICE

Artikelnummer. 18765466
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18765466 10 mg
18746176 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18765466 Leverandør Tocris Bioscience™ Leverandørnr. 7489/10

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Enhancer of 20S proteasome assembly and 20S-mediated protein degradation

TCH-165 is an enhancer of 20S proteasome assembly. TCH-165 regulates the dynamic equilibrium between the 20S and 26S proteasome complexes, favoring 20S-mediated protein degradation. TCH-165 enhances the proteolytic activity of the 20S proteasome by increasing substrate accessibility to the 20S catalytic chamber through 20S gate opening. TCH-165 enhances 20S-mediated degradation of IDPs, α-syn, and tau in vitro, and does not induce the degradation of structured proteins such as GAPDH. TCH-165 enhances c-MYC degradation by the 20S proteasome, which reduces c-MYC levels in vivo, and is effective in bortezomib (Cat. No. 7282) resistant cells. TCH-165 down-regulates c-MYC target genes and inhibits cancer cell proliferation and tumor growth in vivo.
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Ethyl (4 R,5 R)-rel-1-benzyl-5-(4-(benzylamino)phenyl)-2-(4-methoxyphenyl)-4-phenyl-4,5-dihydro-1 H-imidazole-4-carboxylate
CAS 1446350-60-2
Mængde 10 mg
Mål Proteasome Modulators
Molekylær formel C39H37N3O3
Renhed 0.98
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.