Få mere at vide
TCEP-d12 Hydrochloride, TRC
Artikelnummer. 30412414
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30412414 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412414 Leverandør Toronto Research Chemicals Leverandørnr. TRCT01250210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale TCEP-d12 Hydrochloride
CAS 1189959-10-1
Renhedsgrad noter HPLC
Molekylær formel C9H4D12ClO6P
Mængde 10 mg
Synonym 3-Bis(1,1,2,2-tetradeuterio-3-hydroxy-3-oxopropyl)phosphanyl-2,2,3,3-tetradeuteriopropanoic acid hydrochloride
InChI formel InChI=1S/C9H15O6P.ClH/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15;/h1-6H2,(H,10,11)(H,12,13)(H,14,15);1H/i1D2,2D2,3D2,4D2,5D2,6D2;
SMIL [2H]C([2H])(C(O)=O)C([2H])([2H])P(C([2H])([2H])C([2H])([2H])C(O)=O)C([2H])([2H])C([2H])([2H])C(O)=O.Cl
IUPAC navn 3,3',3''-phosphanetriyltris(propanoic-2,2,3,3-d4 acid) hydrochloride
Molekylvægt (g/mol) 298.72
Formel vægt 298.11
Procent renhed >95