Få mere at vide
Tocris Bioscience™ TC-S 7010
Potent and selective Aurora kinase A inhibitor
Brand: Tocris Bioscience™ 5286/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TC-S 7010 is a potent and selective inhibitor of Aurora kinase A (IC50 = 3.4 nM); exhibits 1000-fold selectivity for Aurora kinase A over Aurora kinase B. Displays antiproliferative activity in HCT116 and HT29 cells (IC50 values are 0.19 and 2.9 μM respectively).Tekniske data
| N-(2-Chlorophenyl)-4-[[2-[[4-[2-(4-ethyl-1-piperazinyl)-2-oxoethyl]phenyl]amino]-5-fluoro-4-pyrimidinyl]amino]-benzamide | |
| 1158838-45-9 | |
| 50 mg |