missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ TC-G 1001
GREENER_CHOICE

Artikelnummer. 30001669
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001669 50 mg
30001655 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30001669 Supplier Tocris Bioscience™ Supplier No. 4879/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

GPR35 agonist; more potent than zaprinast (Cat. No. 0947)

GPR35 agonist (pEC50 values are 7.59 and 8.36 for β-arrestin and Gαq-i5 Ca2+ assays respectively). Displays 1000-fold greater potency at human GPR35 receptors relative to mouse and rat GPR35 orthologs in an IP1 accumulation assay. More potent than zaprinast (Cat. No. 0947).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 494191-73-0
Molekylær formel C17H11FN2O3S

Specifications

Kemisk navn eller materiale TC-G 1001
Mængde 50 mg
Formel vægt 342.34
Renhed >98%
Molekylvægt (g/mol) 342.344
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.