Få mere at vide
Tocris Bioscience™ TC-G 1001
GREENER_CHOICE
Artikelnummer. 30001655
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30001655 10 mg
30001669 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30001655 Leverandør Tocris Bioscience™ Leverandørnr. 4879/10
GPR35 agonist; more potent than zaprinast (Cat. No. 0947)
GPR35 agonist (pEC50 values are 7.59 and 8.36 for β-arrestin and Gαq-i5 Ca2+ assays respectively). Displays 1000-fold greater potency at human GPR35 receptors relative to mouse and rat GPR35 orthologs in an IP1 accumulation assay. More potent than zaprinast (Cat. No. 0947).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 494191-73-0
Molekylær formel C17H11FN2O3S
Tekniske data
Kemisk navn eller materiale TC-G 1001
Mængde 10 mg
Formel vægt 342.34
Renhed >98%
Molekylvægt (g/mol) 342.344