missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D10B Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP3-05943AF532
Additional Details : Weight : 0.00970kg
Description
TBC1D10B Polyclonal specifically detects TBC1D10B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
TBC1D10B | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EPI64B, FP2461, TBC1D10B TBC1 domain family, member 10B | |
Produced in rabbits immunized with E. coli-derived Human TBC1D10B fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
26000 | |
Store at 4°C in the dark. | |
IgG |