missing translation for 'onlineSavingsMsg'
Learn More

TB 500, TRC

Product Code. 30421165
Change view
Click to view available options
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30421165 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421165 Supplier Toronto Research Chemicals Supplier No. TRCT0102051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (2S)-2-(((2S,3R)-2-(((2S)-2-(((2S,3R)-2-(((2S)-2-(((2S)-2-(((2S)-2-acetamido-4-methylpentanoyl)amino)-6-aminohexanoyl)amino)-6-aminohexanoyl)amino)-3-hydroxybutanoyl)amino)-4-carboxybutanoyl)amino)-3-hydroxybutanoyl)amino)-5-amino-5-oxopentanoic acid
CAS 885340-08-9
Renhedsgrad noter HPLC
Molekylær formel C38 H68 N10 O14
Mængde 1 mg
Synonym (2S)-2-[[(2S,3R)-2-[[(2S)-2-[[(2S,3R)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-acetamido-4-methylpentanoyl]amino]-6-aminohexanoyl]amino]-6-aminohexanoyl]amino]-3-hydroxybutanoyl]amino]-4-carboxybutanoyl]amino]-3-hydroxybutanoyl]amino]-5-amino-5-oxopentanoic acid, TB 500
InChI formel InChI=1S/C38H68N10O14/c1-19(2)18-27(42-22(5)51)35(58)44-23(10-6-8-16-39)32(55)43-24(11-7-9-17-40)33(56)47-30(20(3)49)36(59)45-25(13-15-29(53)54)34(57)48-31(21(4)50)37(60)46-26(38(61)62)12-14-28(41)52/h19-21,23-27,30-31,49-50H,6-18,39-40H2,1-5H3,(H2,41,52)(H,42,51)(H,43,55)(H,44,58)(H,45,59)(H,46,60)(H,47,56)(H,48,57)(H,53,54)(H,61,62)/t20-,21-,23+,24+,25+,26+,27+,30+,31+/m1/s1
SMIL CC(C)C[C@H](NC(=O)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCC(=O)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CCC(=O)N)C(=O)O
IUPAC navn (2S)-2-[[(2S,3R)-2-[[(2S)-2-[[(2S,3R)-2-[[(2S)-2-[[(2S)-2-[[(2S)-2-acetamido-4-methylpentanoyl]amino]-6-aminohexanoyl]amino]-6-aminohexanoyl]amino]-3-hydroxybutanoyl]amino]-4-carboxybutanoyl]amino]-3-hydroxybutanoyl]amino]-5-amino-5-oxopentanoic acid
Molekylvægt (g/mol) 889.005
Formel vægt 888.492
Procent renhed >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.