Få mere at vide
Tocris Bioscience™ TB 21007
GREENER_CHOICE
Artikelnummer. 18084592
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18084592 50 mg
18035552 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18084592 Leverandør Tocris Bioscience™ Leverandørnr. 2905/50
α5-selective GABAA inverse agonist
GABAA receptor inverse agonist selective for the α5-subtype (Ki values are 1.6, 16, 20 and 20 nM for α5, α2, α1 and α3 subtypes respectively). Brain penetrant; enhances cognitive performance in rats in the delayed matching-to-place morris water maze test following i.p. administration.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 207306-50-1
Molekylær formel C15H17NO2S3
Tekniske data
Kemisk navn eller materiale TB 21007
Mængde 50 mg
Formel vægt 339.5
Renhed >99%
Molekylvægt (g/mol) 339.486