missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tazobactam, MedChemExpress
Beskrivelse
Tazobactam (CL-298741) is a potent β-lactamases inhibitor and penicillin antibiotic. Tazobactam has antibacterial activity. Tazobactam can be used for pneumonia research.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tazobactam |
| Fysisk form | Powder |
| CAS | 89786-04-9 |
| Farve | White |
| Mængde | 200 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C10H12N4O5S |
| Til brug med (applikation) | COVID-19-immunoregulation |
| Synonym | CL-298741 YTR-830H |
| Opløselighedsinformation | DMSO : 50 mg/mL (166.51 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.33 mM; ultrasonic and warming and heat to 60°C) |
| Vis mere |
Research purposes only
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?