Få mere at vide
Tazobactam Diphenylmethyl Ester, Mikromol™
Beskrivelse
Tazobactam Diphenylmethyl Ester
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tazobactam Diphenylmethyl Ester |
| CAS | 89789-07-1 |
| Analyt- eller komponentnavne | Tazobactam Diphenylmethyl Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C23 H22 N4 O5 S |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (2S,3S,5R)-3-Methyl-7-oxo-3-(1H-1,2,3-triazol-1-ylmethyl)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid diphenylmethyl ester 4,4-dioxide, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 3-methyl-7-oxo-3-(1H-1,2,3-triazol-1-ylmethyl)-, diphenylmethyl ester, 4,4-dioxide, [2S-(2α,3β,5α)]-, 1H-1,2,3-Triazole, 4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid deriv., Tazobactam diphenylmethyl ester |
| InChI formel | InChI=1S/C23H22N4O5S/c1-23(15-26-13-12-24-25-26)21(27-18(28)14-19(27)33(23,30)31)22(29)32-20(16-8-4-2-5-9-16)17-10-6-3-7-11-17/h2-13,19-21H,14-15H2,1H3/t19-,21+,23+/m1/s1 |
| SMIL | C[C@]1(Cn2ccnn2)[C@@H](N3[C@@H](CC3=O)S1(=O)=O)C(=O)OC(c4ccccc4)c5ccccc5 |
| IUPAC navn | benzhydryl (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-(triazol-1-ylmethyl)-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| Vis mere |