Få mere at vide
Tazarotenic Acid, TRC
Artikelnummer. 30138643
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138643 5 mg
30132962 25 mg
30005384 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138643 Leverandør Toronto Research Chemicals Leverandørnr. TRCT0100605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 118292-41-4
Molekylær formel C19 H17 N O2 S
Molekylvægt (g/mol) 323.41
Synonym 3-Pyridinecarboxylic acid, 6-[2-(3,4-dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]- (ACI), 2H-1-Benzothiopyran, 3-pyridinecarboxylic acid deriv. (ZCI), 3-Pyridinecarboxylic acid, 6-[(3,4-dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]- (9CI), 6-[2-(3,4-Dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]-3-pyridinecarboxylic acid (ACI), AGN 190299, Tazarotenic acid
IUPAC navn 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylic acid
SMIL CC1(C)CCSc2ccc(cc12)C#Cc3ccc(cn3)C(=O)O
Tekniske data
Kemisk navn eller materiale Tazarotenic acid
Mængde 5 mg
InChI formel InChI=1S/C19H17NO2S/c1-19(2)9-10-23-17-8-4-13(11-16(17)19)3-6-15-7-5-14(12-20-15)18(21)22/h4-5,7-8,11-12H,9-10H2,1-2H3,(H,21,22)
Formel vægt 323.098