Få mere at vide
Tazarotenic Acid Sulfoxide, TRC
Artikelnummer. 30132386
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132386 100 mg
30011582 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132386 Leverandør Toronto Research Chemicals Leverandørnr. TRCT010070100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 603952-64-3
Molekylær formel C19 H17 N O3 S
Molekylvægt (g/mol) 339.41
Synonym 6-[2-(3,4-Dihydro-4,4-dimethyl-1-oxido-2H-1-benzothiopyran-6-yl)ethynyl]-3-pyridinecarboxylic Acid, 6-[(3,4-Dihydro-4,4-dimethyl-1-oxido-2H-1-benzothiopyran-' '¡6-yl)ethynyl]-3-pyridinecarboxylic Acid;
IUPAC navn 6-[2-(4,4-dimethyl-1-oxo-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylic acid
SMIL CC1(C)CCS(=O)c2ccc(cc12)C#Cc3ccc(cn3)C(=O)O
Tekniske data
Kemisk navn eller materiale Tazarotenic Acid Sulfoxide
Mængde 100 mg
InChI formel InChI=1S/C19H17NO3S/c1-19(2)9-10-24(23)17-8-4-13(11-16(17)19)3-6-15-7-5-14(12-20-15)18(21)22/h4-5,7-8,11-12H,9-10H2,1-2H3,(H,21,22)
Formel vægt 339.0929