Learn More
Tazarotene Thiochromane Dimer, TRC
Product Code. 30009601
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009601 100 mg 100mg
30009216 25 mg 25mg
30008141 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009601 Supplier Toronto Research Chemicals Supplier No. TRCT010055100MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1010805-73-8
Molecular Formula C26 H26 S2
Molecular Weight (g/mol) 402.6146
Synonym 6,6'-(1,3-Butadiyne-1,4-diyl)bis[3,4-dihydro-4,4-dimethyl-2H-1-benzothiopyran
IUPAC Name 6-[4-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)buta-1,3-diynyl]-4,4-dimethyl-2,3-dihydrothiochromene
SMILES CC1(C)CCSc2ccc(cc12)C#CC#Cc3ccc4SCCC(C)(C)c4c3
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C26H26S2/c1-25(2)13-15-27-23-11-9-19(17-21(23)25)7-5-6-8-20-10-12-24-22(18-20)26(3,4)14-16-28-24/h9-12,17-18H,13-16H2,1-4H3
Formula Weight 402.1476
Chemical Name or Material Tazarotene Thiochromane Dimer