Få mere at vide
Tazarotene Sulfone, TRC
Artikelnummer. 30136470
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136470 100 mg
30138465 10 mg
30005124 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136470 Leverandør Toronto Research Chemicals Leverandørnr. TRCT010080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1262228-27-2
Molekylær formel C21H21NO4S
Molekylvægt (g/mol) 383.46
Synonym Ethyl 6-((4,4-dimethyl-1,1-dioxidothiochroman-6-yl)ethynyl)nicotinate, Ethyl Ester 6-[2-(3,4-Dihydro-4,4-dimethyl-1,1-dioxido-2H-1-benzothiopyran-6-yl)ethynyl]-3-pyridinecarboxylic Acid, Tazarotene Sulphone;
SMIL CCOC(=O)c1ccc(nc1)C#Cc2ccc3c(c2)C(C)(C)CCS3(=O)=O
Tekniske data
Kemisk navn eller materiale Tazarotene Sulfone
Mængde 100 mg
InChI formel InChI=1S/C21H21NO4S/c1-4-26-20(23)16-7-9-17(22-14-16)8-5-15-6-10-19-18(13-15)21(2,3)11-12-27(19,24)25/h6-7,9-10,13-14H,4,11-12H2,1-3H3