missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ Taxol

Klik for at se tilgængelige muligheder
Mængde:
10 mg
50 mg
Beskrivelse
Taxol, also known as paclitaxel, promotes and stabilizes tubulin polymerization, causing cell cycle arrest. In CCRF-HSB-2 cells, Taxol induces autocatalytic activation of caspase-10, triggering apoptosis. Taxol is an antitumor agent. In a mouse model of bladder cancer Taxol decreases tumor growth. Taxol also prolongs survival of mice transplanted with human ovarian carcinoma xenografts. Taxol improves neurological outcome and magnetic resonance imaging biomarkers after traumatic brain injury in mice.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Taxol |
| Mængde | 10 mg |
| Formel vægt | 853.92 |
| Renhed | >99% |
| Molekylvægt (g/mol) | 853.92 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion