Få mere at vide
Taurolithocholic Acid-d5 Sodium Salt (Major), TRC
Artikelnummer. 30417637
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30417637 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30417637 Leverandør Toronto Research Chemicals Leverandørnr. TRCT00910210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Taurolithocholic Acid-d5 Sodium Salt
CAS 1265476-97-8
Renhedsgrad noter HPLC
Molekylær formel C26H39D5NNaO5S
Mængde 10 mg
Synonym 2-[[(3α,5β)-3-Hydroxy-24-oxocholan-24-yl-2,2,3,4,4-d5]amino]ethanesulfonic acid sodium salt, Taurolithocholic acid-d5 sodium salt, Taurolithocholic acid D5 sodium salt (3α-hydroxy-5β-cholane-2,2,3,4,4-D5), 2-[[(3α,5β)-3-Hydroxy-24-oxo-2,2,3,4,4-d5-cholan-24-yl]amino]ethanesulfonic acid sodium salt, 3α-Hydroxy-2,2,3,4,4-d5-5β-cholanoyltaurine sodium, 3α-Hydroxy-N-(2-sulfoethyl)-5β-cholan-24-amide-2,2,3,4,4-d5 sodium salt, Lithocholyltaurine-d5 sodium
InChI formel InChI=1S/C26H45NO5S.Na/c1-17(4-9-24(29)27-14-15-33(30,31)32)21-7-8-22-20-6-5-18-16-19(28)10-12-25(18,2)23(20)11-13-26(21,22)3;/h17-23,28H,4-16H2,1-3H3,(H,27,29)(H,30,31,32);/q;+1/p-1/t17-,18-,19-,20+,21-,22+,23+,25+,26-;/m1./s1/i10D2,16D2,19D;
SMIL [Na+].[2H]C1([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC[C@@H]2C([2H])([2H])[C@]1([2H])O)[C@H](C)CCC(=O)NCCS(=O)(=O)[O-]
IUPAC navn sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-2,2,3,4,4-pentadeuterio-3-hydroxy-10,13-dimethyl-1,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
Molekylvægt (g/mol) 510.72
Formel vægt 510.3152
Procent renhed >95