Få mere at vide
Taurocholic Acid, TRC
Artikelnummer. 17907254
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17907254 10 mg
17958294 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17907254 Leverandør Toronto Research Chemicals Leverandørnr. TRCT00814510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81-24-3
Molekylær formel C26 H45 N O7 S
Molekylvægt (g/mol) 515.7
Synonym 2-[[(3α,5β,7α,12α)-3,7,12-Trihydroxy-24-oxocholan-24-yl]amino]ethanesulfonic Acid;' '¡3α,7α,12α-Trihydroxy-5β-cholanic Acid-24-taurine, Cholaic Acid, Cholic Acid Taurine Conjugate, Cholyltaurine, N-Choloyltaurine, NSC 25505;
IUPAC navn 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulfonic acid
SMIL O=S(O)(CCNC(CC[C@@H](C)[C@]1([H])[C@]2(C)[C@](CC1)([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@@H]2O)=O)=O
Tekniske data
Kemisk navn eller materiale Taurocholic Acid
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C26H45NO7S/c1-15(4-7-23(31)27-10-11-35(32,33)34)18-5-6-19-24-20(14-22(30)26(18,19)3)25(2)9-8-17(28)12-16(25)13-21(24)29/h15-22,24,28-30H,4-14H2,1-3H3,(H,27,31)(H,32,33,34)/t15-,16+,17-,18-,19+,20+,21-,22+,24+,25+,26-/m1/s1
Formel vægt 515.29
Procent renhed >95%