missing translation for 'onlineSavingsMsg'
Få mere at vide

Taurochenodeoxycholic Acid Sodium Salt, TRC

Artikelnummer. 30133503
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133503 1 g
30137338 500 mg
17923855 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30133503 Leverandør Toronto Research Chemicals Leverandørnr. TRCT0081301G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 6009-98-9
Molekylær formel C26 H44 N O6 S . Na
Molekylvægt (g/mol) 521.69
Synonym Ethanesulfonic acid, 2-[[(3α,5β,7α)-3,7-dihydroxy-24-oxocholan-24-yl]amino]-, sodium salt (1:1), Ethanesulfonic acid, 2-[[(3α,5β,7α)-3,7-dihydroxy-24-oxocholan-24-yl]amino]-, monosodium salt (9CI), Taurine, N-(3α,7α-dihydroxy-5β-cholan-24-oyl)-, monosodium salt (8CI), Taurochenodeoxycholic acid sodium salt (6CI), NSC 681055, Sodium taurochenodeoxycholate, Sodium taurochenodesoxycholate
IUPAC navn sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
SMIL [Na+].C[C@H](CCC(=O)NCCS(=O)(=O)[O-])[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Tekniske data

Kemisk navn eller materiale Taurochenodeoxycholic Acid Sodium Salt
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C26H45NO6S.Na/c1-16(4-7-23(30)27-12-13-34(31,32)33)19-5-6-20-24-21(9-11-26(19,20)3)25(2)10-8-18(28)14-17(25)15-22(24)29;/h16-22,24,28-29H,4-15H2,1-3H3,(H,27,30)(H,31,32,33);/q;+1/p-1/t16-,17+,18-,19-,20+,21+,22-,24+,25+,26-;/m1./s1
Formel vægt 521.2787
Procent renhed >95%
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.