Få mere at vide
Tau-(Methyl-d3)-L-Histidine, TRC
Artikelnummer. 30455361
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30455361 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455361 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7793530.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tau-(Methyl-d3)-L-Histidine
CAS 91037-48-8
Molekylær formel C7 D3 H8 N3 O2
Mængde 0.5 mg
Synonym 1-(Methyl-d3)-L-histidine
InChI formel InChI=1S/C7H11N3O2/c1-10-3-5(9-4-10)2-6(8)7(11)12/h3-4,6H,2,8H2,1H3,(H,11,12)/t6-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])n1cnc(C[C@H](N)C(=O)O)c1
IUPAC navn (2S)-2-amino-3-[1-(trideuteriomethyl)imidazol-4-yl]propanoic acid
Molekylvægt (g/mol) 172.2
Formel vægt 172.104