Få mere at vide
Tocris Bioscience™ TASP 0390325
GREENER_CHOICE
Potent and selective V1B receptor antagonist; orally bioavailable
Brand:  Tocris Bioscience™ 5729/50
Encompass
Produktkode 15787076
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
TASP 0390325 is a potent and selective V1B receptor antagonist (IC50 = 2.22 nM for rat pituitary receptor). Exhibits no significant affinity for 85 other receptors, ion channels and transporters at 10 μM, including V1A and V2. Increases ACTH levels in rats. Orally bioavailable. Antidepressant and anxiolytic.
Tekniske data
Tekniske data
Potent and Selective V1B Receptor Antagonist, Orally Bioavailable
1642187-96-9
554.44g/mol
Vasopressin Receptor Antagonists
Vasopressin Receptor
C25H29ClFN5O4.HCl
Store at -20°C
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only