missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ TAS2R46 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15806193
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15806193 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15806193 Leverandør Invitrogen™ Leverandørnr. PA567772

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Most taste cells may be activated by a limited number of bitter compounds; individual taste cells can discriminate among bitter stimuli.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen TAS2R46
Applikationer Western Blot
Klassifikation Polyclonal
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAS2R46
Genadgangsnr. P59540
Gene Alias MGC134865; T2R46; T2R54; TAS2R46; TAS2R47; taste 2 receptor member 46; taste receptor type 2 member 46; Taste receptor type 2 member 54; taste receptor, type 2, member 46
Gensymboler TAS2R46
Værtsarter Rabbit
Immunogen Synthesized peptide derived from human TAS2R46.
Oprensningsmetode Affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 259292
Målarter Human, Mouse, Rat
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.