Få mere at vide
Tartaric Acid Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tartaric Acid Methyl Ester |
| CAS | 3333-46-8 |
| Molekylær formel | C5H8O6 |
| Mængde | 100 mg |
| Synonym | (2R,3R)-2,3-Dihydroxy-butanedioic Acid Monomethyl Ester, 2,3-Dihydroxy-[R-(R*,R*)]-butanedioic Acid Monomethyl Ester, Tartaric Acid Monomethyl Ester, (+)-Methyl Hydrogen Tartrate, (2R,3R)-2,3-Dihydroxybutanedioic Acid Monomethyl Ester |
| InChI formel | InChI=1S/C5H8O6/c1-11-5(10)3(7)2(6)4(8)9/h2-3,6-7H,1H3,(H,8,9)/t2-,3-/m1/s1 |
| SMIL | COC([C@H](O)[C@@H](O)C(O)=O)=O |
| IUPAC navn | (2R,3R)-2,3-dihydroxy-4-methoxy-4-oxobutanoic acid |
| Molekylvægt (g/mol) | 164.11 |
| Formel vægt | 164.0321 |