Få mere at vide
Thermo Scientific™ TARGET DP 350μL insert vials
Artikelnummer. 15348046
Artikelnummer. 15348046
Brand: Thermo Scientific™ CERT500073LV
Use certified clean, low particle, low background vials for your high sensitivity LC/MS and GC/MS applications.
  • 100 vials and 100 caps with pre-assembled septa
  • Reusable two-compartment trays protect vials and closures while keeping matching supplies together
  • Clear trays make it easy to keep track of available supplies without opening containers