Få mere at vide
Taranabant racemate, MedChemExpress
Artikelnummer. 16492788
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
Pakningsstørrelse:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16492788 10 mg 10mg
16435537 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16492788 Leverandør Medchem Express Leverandørnr. HY10013A
MedChemExpress Taranabant racemate (MK-0364 racemate) is an antagonist and/or inverse agonist of the Cannabinoid-1 (CB1) receptor extracted from patent WO 2004048317 A1.
Taranabant racemate (MK-0364 racemate) is an antagonist and/or inverse agonist of the Cannabinoid-1 (CB1) receptor extracted from patent WO 2004048317 A1.
Tekniske data
Kemisk navn eller materiale Taranabant racemate
Fysisk form Solid
CAS 701977-00-6
Farve White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C27H25ClF3N3O2
Til brug med (applikation) Neuroscience-Neuromodulation
Synonym MK-0364 racemate
Opløselighedsinformation DMSO : 100 mg/mL (193.82 mM; Need ultrasonic)
SMIL CC(OC1=NC=C(C(F)(F)F)C=C1)(C)C(N[C@H](C)[C@@H](C2=CC=CC(C#N)=C2)CC3=CC=C(Cl)C=C3)=O
Molekylvægt (g/mol) 515.95
Formel vægt 515.95
Procent renhed 99.58%