Learn More
TAPT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560025
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EEKLSNPPAT CTPGKPSSKS QNKCKPSQGL STEENLSASI TKQPIHQKEN IIPLLVTSNS DQFLTTPDGD EKDITQDNSE LKHRSSKKD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
This gene encodes a highly conserved, putative transmembrane protein. A mutation in the mouse ortholog of this gene results in homeotic, posterior-to-anterior transformations of the axial skeleton which are similar to the phenotype of mouse homeobox C8 gene mutants. This gene is proposed to function downstream of homeobox C8 to transduce extracellular patterning information during axial skeleton development. An alternatively spliced transcript variant encoding a substantially different isoform has been described, but its biological validity has not been determined.Specifications
TAPT1 | |
Polyclonal | |
Unconjugated | |
TAPT1 | |
4932414K18Rik; CMVFR; Cytomegalovirus partial fusion receptor; OCLSBG; TAPT1; transmembrane anterior posterior transformation 1; transmembrane anterior posterior transformation protein 1; Transmembrane anterior posterior transformation protein 1 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
202018 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NXT6 | |
TAPT1 | |
Recombinant protein corresponding to Human TAPT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only