Få mere at vide
TAPI-1, MedChemExpress
Artikelnummer. 16459647
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16459647 5 mg
16421629 1 mg
16407108 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16459647 Leverandør Medchem Express Leverandørnr. HY16657
MedChemExpress TAPI-1 is a TACE (ADAM17) inhibitor and blocks the shedding of several cell surface proteins. TAPI-1 is also a metalloproteinase (MMP) inhibitor.
TAPI-1 is a TACE (ADAM17) inhibitor and blocks the shedding of several cell surface proteins. TAPI-1 is also a metalloproteinase (MMP) inhibitor.
Tekniske data
Kemisk navn eller materiale TAPI-1
Fysisk form Powder
CAS 163847-77-6
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C26H37N5O5
Til brug med (applikation) Cancer-Kinase/protease
Opløselighedsinformation DMSO : ≥ 31 mg/mL (62.05 mM)
SMIL O=C(N[C@@H](CC1=CC=C2C=CC=CC2=C1)C(N[C@@H](C)C(NCCN)=O)=O)[C@H](CC(C)C)CC(NO)=O
Molekylvægt (g/mol) 499.6
Formel vægt 499.6
Procent renhed 99.0%