missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TANK Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0386
Additional Details : Weight : 0.01000kg
Description
TANK Synthetic Peptide, PEP-0386, from Invitrogen.
This peptide corresponds to 14 amino acids near the amino terminus of human TANK.PEP-0386 can be used as a blocking peptide with polyclonal antibody PA5-20271.Specifications
10010 | |
purified | |
Human TANK | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q92844 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TANK | |
0.02% sodium azide | |
50 μg | |
PBS |