Få mere at vide
Tamsulosin EP D as Hydrochloride, Mikromol™
Artikelnummer. 30461928
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30461928 4 x 25 mg
30049388 25 mg
30051924 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30461928 Leverandør Mikromol Leverandørnr. MM0938.0500254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tamsulosin EP D as Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Methoxy-5-[(2R)-2-[(2-(2-methoxyphenoxy)ethyl)amino]propyl]benzenesulfonamide Hydrochloride
CAS 1798904-07-0
Analyt- eller komponentnavne 2-Methoxy-5-[(2R)-2-[(2-(2-methoxyphenoxy)ethyl)amino]propyl]benzenesulfonamide Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C19 H26 N2 O5 S . H Cl
Certificeringer/compliance ISO 17025
Synonym 2-Methoxy-5-[(2R)-2-[[2-(2-methoxyphenoxy)ethyl]amino]propyl]benzenesulfonamide Hydrochloride, Benzenesulfonamide, 2-methoxy-5-[(2R)-2-[[2-(2-methoxyphenoxy)ethyl]amino]propyl]-, hydrochloride (1:1), (R)-2'-O-Desethyl-2'-O-desmethyl Tamsulosin hydrochloride
InChI formel InChI=1S/C19H26N2O5S.ClH/c1-14(21-10-11-26-17-7-5-4-6-16(17)24-2)12-15-8-9-18(25-3)19(13-15)27(20,22)23;/h4-9,13-14,21H,10-12H2,1-3H3,(H2,20,22,23);1H/t14-;/m1./s1
Løsningstype Neat
SMIL Cl.COc1ccccc1OCCN[C@H](C)Cc2ccc(OC)c(c2)S(=O)(=O)N
IUPAC navn 2-methoxy-5-[(2R)-2-[2-(2-methoxyphenoxy)ethylamino]propyl]benzenesulfonamide;hydrochloride
Molekylvægt (g/mol) 430.94
Mængde 4 x 25 mg
Formel vægt 430.936 g/mol