Få mere at vide
Tamoxifen EP Impurity G, Mikromol™
Artikelnummer. 30457736
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30457736 4 x 25 mg
30049097 100 mg
30049808 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30457736 Leverandør Mikromol Leverandørnr. MM0053.0700254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tamoxifen EP Impurity G, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2RS)-1-[4-[2-(Dimethylamino)-ethoxy]phenyl]-2-phenylbutan-1-one
CAS 68047-07-4
Analyt- eller komponentnavne (2RS)-1-[4-[2-(Dimethylamino)-ethoxy]phenyl]-2-phenylbutan-1-one
Forsendelsestilstand Room Temperature
Molekylær formel C20 H25 N O2
Certificeringer/compliance ISO 17025
Synonym Tamoxifen Citrate Imp. G (EP), Tamoxifen Imp. G (EP), (2RS)-1-[4-[2-(Dimethylamino)-ethoxy]phenyl]-2-phenylbutan-1-one, Tamoxifen Citrate Impurity G, Tamoxifen Impurity G
InChI formel InChI=1S/C20H25NO2/c1-4-19(16-8-6-5-7-9-16)20(22)17-10-12-18(13-11-17)23-15-14-21(2)3/h5-13,19H,4,14-15H2,1-3H3
Løsningstype Neat
SMIL CCC(C(=O)c1ccc(OCCN(C)C)cc1)c2ccccc2
IUPAC navn 1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenylbutan-1-one
Molekylvægt (g/mol) 311.42
Mængde 4 x 25 mg
Formel vægt 311.1885 g/mol