Få mere at vide
Tamoxifen-d5 (ethyl-d5), CDN
Artikelnummer. 30312909
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312909 0.005 g
30321966 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312909 Leverandør CDN Leverandørnr. CDND66370.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tamoxifen-d5
CAS 157698-32-3
Molekylær formel C26 D5 H24 N O
Mængde 0.005 g
Synonym Ethanamine, 2-[4-[(1 Z)-1,2-diphenyl-1-buten-1-yl-3,3,4,4,4-d5]phenoxy]-N,N-dimethyl-, Ethanamine, 2-[4-(1,2-diphenyl-1-butenyl-3,3,4,4,4-d5)phenoxy]-N,N-dimethyl-, (Z)-, Ethanamine, 2-[4-[(1 Z)-1,2-diphenyl-1-butenyl-3,3,4,4,4-d5]phenoxy]-N,N-dimethyl- (9 CI), 2-[4-[(1 Z)-1,2-Diphenyl-1-buten-1-yl-3,3,4,4,4-d5]phenoxy]-N,N-dimethylethanamine, Tamoxifen-d5, Z-Tamoxifen-d5, trans-Tamoxifen-d5, Tamoxifen-D5 (ethyl-D5)
InChI formel InChI=1 S/C26H29NO/c1-4-25(21-11-7-5-8-12-21)26(22-13-9-6-10-14-22)23-15-17-24(18-16-23)28-20-19-27(2)3/h5-18 H,4,19-20H2,1-3H3/b26-25-/i1D3,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])\C(=C(/c1ccccc1)\c2ccc(OCCN(C)C)cc2)\c3ccccc3
IUPAC navn N,N-dimethyl-2-[4-[(Z)-3,3,4,4,4-pentadeuterio-1,2-diphenylbut-1-enyl]phenoxy]ethanamine
Molekylvægt (g/mol) 376.545
Formel vægt 376.256 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity