Få mere at vide
MedChemExpress Taltobulin
Artikelnummer. 16419217
Encompass_Preferred
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
16419217 5 mg
1 options
Artikelnummer. 16419217 Leverandør MedChemExpress Leverandørnr. HY15584
MedChemExpress Taltobulin (HTI-286), a synthetic analogue of the tripeptide hemiasterlin, is a potent antimicrotubule agent that circumvents P-glycoprotein-mediated resistance in vitro and in vivo. Taltobulin inhibits the polymerization of purified tubulin, disrupts microtubule organization in cells, and induces mitotic arrest, as well as apoptosis.
Tekniske data
Renhed 98.07%
CAS 228266-40-8
Indhold og opbevaring Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Til brug med (applikation) Cancer-programmed cell death
Molekylvægt (g/mol) 473.65 g/mol
Produkttype Reference compound
Mængde 5 mg
Forsendelsestilstand Room temperature
Opløselighed DMSO : ≥ 100 mg/mL (211.13 mM)
Synonym HTI-286∣ SPA-110