missing translation for 'onlineSavingsMsg'
Learn More

Talniflumat, TRC

Product Code. 30422511
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30422511 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422511 Supplier Toronto Research Chemicals Supplier No. TRCT00557510MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Talniflumate
CAS 66898-62-2
Molekylær formel C21H13F3N2O4
Mængde 10 mg
Synonym BA 7602-06, Somalgen, 2-[[3-(Trifluoromethyl)phenyl]amino]-','1,3-dihydro-3-oxo-1-isobenzofuranyl-3-pyridinecarboxylic Acid Ester
InChI formel InChI=1S/C21H13F3N2O4/c22-21(23,24)12-5-3-6-13(11-12)26-17-16(9-4-10-25-17)19(28)30-20-15-8-2-1-7-14(15)18(27)29-20/h1-11,20H,(H,25,26)
SMIL FC(F)(F)c1cccc(Nc2ncccc2C(=O)OC3OC(=O)c4ccccc34)c1
IUPAC navn (3-oxo-1H-2-benzofuran-1-yl) 2-[3-(trifluoromethyl)anilino]pyridin-3-carboxylat
Molekylvægt (g/mol) 414.33
Formel vægt 414.0827
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.