Få mere at vide
Talnetant, MedChemExpress
Artikelnummer. 16492227
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
Pakningsstørrelse:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16492227 50 mg 50mg
16406098 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16492227 Leverandør Medchem Express Leverandørnr. HY14552
MedChemExpress Talnetant (SB 223412) is a potent and selective NK3 receptor antagonist (ki=1.4 nM, hNK-3-CHO); 100-fold selective for the hNK-3 versus hNK-2 receptor, with no affinity for the hNK-1 at concentrations up to 100 uM.
Talnetant (SB 223412) is a potent and selective NK3 receptor antagonist (ki=1.4 nM, hNK-3-CHO)
Tekniske data
Kemisk navn eller materiale Talnetant
CAS 174636-32-9
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C25H22N2O2
Mængde 50 mg
Synonym SB 223412
Opløselighedsinformation DMSO : ≥ 100 mg/mL (261.47 mM)
SMIL OC1=C(C(N[C@H](C2=CC=CC=C2)CC)=O)C3=C(N=C1C4=CC=CC=C4)C=CC=C3
Molekylvægt (g/mol) 382.45
Formel vægt 382.45
Procent renhed 99.15%
Grad Research
Fysisk form Solid
Til brug med (applikation) Neuroscience-Neuromodulation