Learn More
Witeg™ Tall Form Glass Beakers
Product Code. 10180650
Change view
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Pack of 10
Product Code. Capacity (Metric) Unit Size
10180650 25 mL Pack of 10
1 options
Product Code. 10180650 Supplier Witeg™ Supplier No. 5 502 025 S
  • ONLINE EXCLUSIVE PRICE
    108.44 DKK
    99.37 DKK / Pakke med 10
    Spare 9.07 DKK
    8% Slukket
View all promotions
Forventet forsendelse: 14-01-2027
With graduations for accurate and precise filling and dispensing.
  • Tall form
  • Acc. to DIN 12331, ISO 3819
  • Manufactured from DURAN™ borosilicate glass 3.3
  • With spout for easy pouring
Specifications
Color Transparent
Capacity (Metric) 25 mL
Material Borosilicate Glass
Includes Spout
Graduated Yes
Certifications/Compliance ISO 3819, DIN 12331
Style Tall Form