missing translation for 'onlineSavingsMsg'
Få mere at vide

(+)-Talarozole, MedChemExpress

Artikelnummer. 16471439
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
5 mg
10 mg
25 mg
50 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16471439 1 mg
16458777 5 mg
16456178 10 mg
16422317 50 mg
16403446 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16471439 Leverandør Medchem Express Leverandørnr. HY14802C

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress (+)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1.

(+)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1.

Tekniske data

Kemisk navn eller materiale (+)-Talarozole
Fysisk form Solid
CAS 201410-66-4
Farve Off-White
Mængde 1 mg
Renhedsgrad noter Research
Molekylær formel C21H23N5S
Til brug med (applikation) Neuroscience-Neuromodulation
Opløselighedsinformation DMSO : ≥ 72 mg/mL (190.72 mM)
SMIL CCC(CC)C(C1=CC=C(NC2=NC3=CC=CC=C3S2)C=C1)N4N=CN=C4.[(+)].[Rotation]
Molekylvægt (g/mol) 377.51
Formel vægt 377.51
Procent renhed 99.28%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.