missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talabostat mesylate, MedChemExpress

Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Antineoplastic and hematopoiesis- stimulating activities.
Specifications
Specifications
Chemical Name or Material | Talabostat mesylate |
CAS | 150080-09-4 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C10H23BN2O6S |
Quantity | 50 mg |
Synonym | Val-boroPro mesylatePT100 mesylate |
Solubility Information | H2O : 250 mg/mL (805.98 mM; Need ultrasonic) ∣DMSO : ≥ 40 mg/mL (128.96 mM) |
SMILES | CC(C)[C@H](N)C(N1[C@H](B(O)O)CCC1)=O.CS(=O)(O)=O |
Molecular Weight (g/mol) | 310.18 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Talabostat mesylate, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction