Learn More
Duran™ Tail Stopcock
Brand:  Duran™ 286037805
Product Code. 11670039
  • 10395.00 DKK / Pack of 50
Only null left
Estimated Shipment: 10-07-2025
Specifications
Duran Tail Stopcock
2.5 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Duran™ Tail Stopcock > Plug Bore: 2.5mm