missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tafazzin/TAZ Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB600-811
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Tafazzin/TAZ Polyclonal specifically detects Tafazzin/TAZ in Human samples. It is validated for Western Blot, Peptide ELISA.
Tekniske data
| Tafazzin/TAZ | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| BTHSEFE, cardiomyopathy, dilated 3A (X-linked), CMD3A, EFE2FLJ27390, G4.5endocardial fibroelastosis 2, LVNCX, Protein G4.5, tafazzin, Taz1, XAP-2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize isoforms 1to 4 (NP_000107.1; NP_851828.1; NP_851829.1; NP_851830.1, respectively). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 10 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_000107.1 | |
| TAFAZZIN | |
| Peptide with sequence C-HLKTQAEQLHNH corresponding to C-Terminus according to NP_000107.1, NP_851828.1, NP_851829.1, NP_851830.1. | |
| 0.1 mg | |
| Transcription Factors and Regulators | |
| 6901 | |
| Human | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion