missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
TAF12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03296-100UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
TAF12 Polyclonal specifically detects TAF12 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.
Tekniske data
| TAF12 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6883 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, KnockDown | |
| Unconjugated | |
| 20kDa, TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF15, TAF2J, TAFII-20/TAFII-15, TAFII20TAFII20/TAFII15, TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD, Transcription initiation factor TFIID 20/15 kDa subunits, transcription initiation factor TFIID subunit 12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-161 of human TAF12 (NP_001128690.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion