missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ TAAR6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567755
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Predicted to react with Mouse and Rat samples.
Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood. Expressed at low abundance in various brain tissues, as well as in fetal liver, but not in the cerebellum or placenta. In the brain, comparable levels of expression in basal ganglia, frontal cortex, substantia nigra, amygdala and hippocampus, highest expression in hippocampus and lowest expression in basal ganglia.
Tekniske data
| TAAR6 | |
| Polyclonal | |
| Unconjugated | |
| TAAR6 | |
| Gm228; mTaar6; RP11-295F4.3; TA4; TAAR6; TAR4; taR-4; TAR6; taR-6; trace amine associated receptor 6; trace amine receptor 4; trace amine receptor 6; trace amine-associated receptor 6; trace-amine-associated receptor 6; TRAR4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 215855, 294124, 319100 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5QD13, Q923Y5, Q96RI8 | |
| TAAR6 | |
| Synthesized peptide derived from human TAAR6. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion