Beskrivelse
- T-Mop covers with snap fasteners in a range of fabrics
- Rounded covered sponges easily roll down walls and soft walls
- Easily fits in awkward corners, tight spaces and under equipment
- Autoclavable
Tekniske data
Tekniske data
| Produkttype | Mop Cover |
| Farve | White |
| Sterilitet | Non-sterile |
| Autoklaverbar | Autoclavable |
| Engangs | Reusable |
| Farvekodet | No |
| Materiale | Microfiber Polyester |