Få mere at vide
(T-4)-Triphenyl(P,P,P-triphenylphosphine imidato-κN)-phosphorus Diphosphate, TRC
Artikelnummer. 30449393
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30449393 100 mg
1 options
Artikelnummer. 30449393 Leverandør Toronto Research Chemicals Leverandørnr. TRCT809325100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (T-4)-Triphenyl(P,P,P-triphenylphosphine imidato-kN)-phosphorus Diphosphate
CAS 1927003-40-4
Molekylær formel C36H34NO7P4+
Mængde 100 mg
Synonym Phosphorus(1+)','triphenyl(P,P,P-triphenylphosphine imidato-κN)-','(T-4)-','diphosphate (1:1) (ACI), (T-4)-Triphenyl(P,P,P-triphenylphosphine imidato-κN)phosphorus diphosphate, Bis(triphenylphosphanylidene)ammonium diphosphate
InChI formel InChI=1S/C36H30NP2.H4O7P2/c1-7-19-31(20-8-1)38(32-21-9-2-10-22-32,33-23-11-3-12-24-33)37-39(34-25-13-4-14-26-34,35-27-15-5-16-28-35)36-29-17-6-18-30-36;1-8(2,3)7-9(4,5)6/h1-30H;(H2,1,2,3)(H2,4,5,6)/q+1;/p-1
SMIL OP(=O)(O)OP(=O)(O)O.c1ccc(cc1)P(=[N+]=P(c2ccccc2)(c3ccccc3)c4ccccc4)(c5ccccc5)c6ccccc6
IUPAC navn bis(triphenyl-λ^{5}-phosphanylidene)azanium;phosphono hydrogen phosphate
Molekylvægt (g/mol) 716.6
Formel vægt 715.1207